Learn More
METTLER TOLEDO Standard MA Analytical Balance
Catalog No. 01804254
Change view
Click to view available options
Capacity:
62 g, 152 g
52 g
92 g
120 g
220 g
Model:
MA104
MA155DU
MA204
MA54
MA55
MA95
Readability:
0.01 mg, 0.1 mg
0.01 mg
0.1 mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Model Readability
01804254 52 g MA54 0.1 mg
01804258 52 g MA55 0.01 mg
01804256 62 g, 152 g MA155DU 0.01 mg, 0.1 mg
01804252 120 g MA104 0.1 mg
01804250 220 g MA204 0.1 mg
01804212 92 g MA95 0.01 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01804254 Supplier METTLER TOLEDO Supplier No. 30697385
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
View all promotions
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
METTLER TOLEDO Standard MA Analytical Balances are highly accurate, efficient, simple to operate and designed with robust construction for proven long lasting performance.
The LCD hybrid touchscreen is simple to operate and will increase efficiency of your daily tasks with 9 built-in applications and statistical data analysis. Built with a metal base, PBT top housing and overload protection to defend against rough handling in any weighing environment, ensuring quality performance for years to come.
MA Analytical Balances include:
  • Internal adjustment
  • LCD hybrid touchscreen with onscreen instructions
  • 9 built-in applications
  • Statistical data analysis
  • Front level indicator
  • Metal base, PBT top housing
  • Overload protection
  • 2 interfaces: USB-A, RS232
  • Bluetooth option
See how it ranks among our balance lines:
  • XPR Excellence Analytical Balances
  • XPR-E Essential Analytical Balances
  • MX Advanced Analytical Balances
  • MR Advanced Analytical Balances
  • MA Standard Analytical Balances
  • FE Standard Analytical Balances
  • LA Standard Analytical Balances
Specifications
Connection Type USB-A, RS232
Capacity 52 g
Display Type Touchscreen
Readability 0.1 mg
Material Metal, PBT
Model MA54
Stabilization Time 2 sec.
Type Analytical Balance
Electrical Requirements 100/240 V, 50/60 Hz
Legal for Trade No