missing translation for 'onlineSavingsMsg'
Learn More

Stanbio™ Hema-Screen™ Lab Pack

Catalog No. SB1290100
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB1290100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB1290100 Supplier Stanbio™ Supplier No. 1290100
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to collect specimens and perform assay in laboratory

  • Provides results in 2 minutes

For in-vitro diagnostic use only

Specifications

Content And Storage Room Temperature
Type Hema Screen™ Lab Pack
For Use With (Application) For in-vitro diagnostic use only
Includes Hema Screen™ developing solution, patient instructions, Wooden applicator and technical insert
Quantity 100 Tests
Test Time 2 min.