missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Stainless Steel Syringe Needles

Long needles feature a female luer lock connector and beveled non-coring tip, designed for use with solvent retrieval.

$22.08 - $24.08

Specifications

Material Stainless Steel
For Use With (Application) Solvent Retrieval
Latex Free Yes
Product Type Syringe Needle
Disposable Reusable
View More Specs

Products 6
Catalog Number Mfr. No. Diameter (Metric) Outer Needle Length (Metric) Needle Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Needle Length (Metric) Needle Price Quantity  
31502325
View Documents
Synthware™
S386150
0.60 mm 150 mm
Each for $22.08
Only null left
 
31502326
View Documents
Synthware™
S386200
0.60 mm 200 mm
Each for $22.68
Only null left
 
31502327
View Documents
Synthware™
S386300
0.60 mm 300 mm
Each for $24.08
Only null left
 
31502328
View Documents
Synthware™
S389150
0.90 mm 150 mm
Each for $22.08
Only null left
 
31502329
View Documents
Synthware™
S389200
0.90 mm 200 mm
Each for $22.68
Only null left
 
31502330
View Documents
Synthware™
S389300
0.90 mm 300 mm
Each for $24.08
Only null left
 
Description

Description

  • Long, stainless steel syringe needles
  • Female luer lock connector
  • Beveled non-coring tip
  • Compatible with both luer slip, and luer lock syringes
  • For use with solvent retrieval
Specifications

Specifications

Stainless Steel
Yes
Reusable
Luer Lock/Luer Slip Syringes
No
Solvent Retrieval
Syringe Needle
Bevel
Luer Lock
Non-sterile
Videos
SDS
Documents

Documents