missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel Spheres, 4mm (0.16 in.) dia., Thermo Scientific Chemicals

Catalog No. AA42021KN
Change view
Click to view available options
Quantity:
100 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42021KN 100 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42021KN Supplier Thermo Scientific Chemicals Supplier No. 042021KN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt%

Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Stainless Steel
Appearance Spheres
Quantity 100 Pc.
Form Sphere
Molecular Formula Fe:Cr:Ni:Mo; 67.5:17:13:12.5 wt%
MDL Number MFCD00148499
Solubility Information Insoluble in water.

RUO – Research Use Only