missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel Spheres, 2mm (0.08 in ) dia., Thermo Scientific Chemicals

Catalog No. AA42019KN
Change view
Click to view available options
Quantity:
100 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42019KN 100 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42019KN Supplier Thermo Scientific Chemicals Supplier No. 042019KN

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Stainless Steel
Appearance Spheres
Quantity 100 Pc.
Form Sphere
Molecular Formula Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
MDL Number MFCD00148499

RUO – Research Use Only