Learn More
Fisherbrand™ Stainless Steel Lab Scoop, 12 Count
Catalog No. 01189170
Change view
Click to view available options
Quantity:
12 Pk.
Catalog No. Quantity
01189170 12 Pk.
1 options
Catalog No. 01189170 Supplier Fisherbrand™ Supplier No. 10401287
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These Stainless Steel Lab Scoops are durable and autoclavable and can be used in a variety of applications. Sold in quantity of 12.
  • Made From ASTM Standard (899-94) Series 400 Stainless Steel
  • Dig out materials, core, and stir easily
  • Minimizes spilling in handling small amounts of material
  • Very versatile
  • One pointed end, one blunt end
  • Will not rust
Specifications
Quantity 12 Pk.
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable
Product Type Scoop
Blade Material Stainless Steel
Material Stainless Steel