Learn More
Fisherbrand™ Stainless-Steel Freezer Racks
Catalog No. 03395604
Change view
Click to view available options
For Use With (Equipment):
2 in. Upright Freezer Boxes
Shelves:
16 (4 x 4 array)
Catalog No. For Use With (Equipment) Shelves
03395604 2 in. Upright Freezer Boxes 16 (4 x 4 array)
1 options
Catalog No. 03395604 Supplier Fisherbrand™ Supplier No. 0439814
Organize and optimize the freezers in the lab
Make it easy to track samples by improving organization in your freezers. These racks enable you to store more samples, eliminate dead space and minimize frost buildup. Fit all standard types and sizes of upright and chest freezers, regardless of manufacturer.
  • Store a variety of sample containers, including boxes, plates, and tubes
  • Sturdy, rust-resistant stainless steel
  • Rack handles at both ends for freezers with right- or left-handed doors
  • Locking rod on chest freezer racks keeps boxes (available separately) securely in place
For more styles and sizes, visit www.fishersci.com.
Specifications
For Use With (Equipment) 2 in. Upright Freezer Boxes
Shelves 16 (4 x 4 array)