missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel Foil, 0.2mm (0.008 in.) thick, Thermo Scientific Chemicals

Catalog No. AA42255GH
Change view
Click to view available options
Quantity:
100 x 100 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42255GH 100 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42255GH Supplier Thermo Scientific Chemicals Supplier No. 042255GH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%

It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Color Silver
Chemical Name or Material Stainless Steel
Name Note Type 304
Appearance Silver-colored foil
Quantity 100 x 100 mm
Odor Odorless
Form Foil
Molecular Formula Fe:Cr:Ni; 70:19:11 wt%
MDL Number MFCD00148499
Solubility Information Insoluble in water.

RUO – Research Use Only