missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Stainless Steel Beaker Tongs

With rubber coated tips, useful for grasping beakers with 50 - 2,000 mL capacities

Supplier:  Eisco™ BKTGSS

Catalog No. S27482


Only null left

Description

Description

  • High quality stainless steel beaker tongs
  • Curved, rubber coated tips for slip free grip
  • Rust and corrosion resistant
Specifications

Specifications

10 in.
Grasping Beakers with 50 - 2,000 mL Capacities
Stainless Steel
Curved, Rubber Coated
Beaker Tongs
254 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions