The dish is made from polymethylpentene "glass clear" for use with our staining rack (Cat# 70321-10) shown above. It comes with two lids; one lid should be used when the rack is not inserted.
Numéro de catalogue.
5028691
Fournisseur
Electron Microscopy SciencesCode fournisseur
7032120
The dish is made from polymethylpentene "glass clear" for use with our staining rack (Cat# 70321-10) shown above. It comes with two lids; one lid should be used when the rack is not inserted.