missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSX1 Monoclonal Antibody (OTI2F8)
GREENER_CHOICE

Numéro de catalogue. PIMA525511
Modifier l'affichage
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIMA525511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIMA525511 Fournisseur Invitrogen™ Code fournisseur MA525511

Mouse Monoclonal Antibody

SSX1 Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

Could act as a modulator of transcription.

Spécifications

Antigen SSX1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2F8
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SSX1
Gene Accession No. Q16384
Gene Alias Cancer/testis antigen 5.1; cancer/testis antigen family 5, member 1; CT5.1; CT5.2; CT51; Protein SSX1; sarcoma, synovial, X-chromosome-related 1; SSRC; SSX family member 1; SSX1; SSX2; SSX2A; SSX2B; SSX3; Synovial sarcoma, X breakpoint 1; X breakpoint 2
Gene Symbols SSX1
Host Species Mouse
Immunogen Full length human recombinant protein of SSX1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6756
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats