Learn More
BeLLCo Glass Stainless steel curved harvesting tube for 8l round bottom omni vessels. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better inside and out electropolish finish. Size: 8l Dimensions: 1/2"" x .430 x 20""
Numéro de catalogue. 501944946
Encompass
Stainless steel curved harvesting tube for 8l round bottom omni vessels. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better inside and out electropolish finish. Size: 8l Dimensions: 1/2" x .430 x 20"
Numéro de catalogue. 501944946 Fournisseur BeLLCo Glass Code fournisseur 196491208
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Stainless steel curved harvesting tube for 8l round bottom omni vessels. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better inside and out electropolish finish. Size: 8l Dimensions: 1/2" x .430 x 20"