Learn More
Chemglass Inc Dish Only, 100 x 65mm, Oil Bath, Stainless Steel
Numéro de catalogue. CG1100D01
Encompass
Dish Only, 100 x 65mm, Oil Bath, Stainless Steel. Stainless steel dish with lid for use as an oil bath when used in conjunction with a hot plate stirrer having a thermocouple connection. Available in three sizes, complete with lids.
Numéro de catalogue. CG1100D01 Fournisseur Chemglass Inc Code fournisseur CG1100D01
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Dish Only, 100 x 65mm, Oil Bath, Stainless Steel. Stainless steel dish with lid for use as an oil bath when used in conjunction with a hot plate stirrer having a thermocouple connection. Available in three sizes, complete with lids.