MW 437.94 g/mol. Potent REV-ERB/ agonist (IC values are 670 and 800 nM for Rev-ErbA and Rev-ErbA-dependent repressor activity respectively). Modulates circadian behavior, shows antiobesogenic and anti-inflammatory effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement