missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SR 16584

Catalog No. 442410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
442410 10 mg
442450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 442410 Supplier Tocris Bioscience™ Supplier No. 4424/10

Selective α3β4 nAChR antagonist

Selective α3β4 nAChR antagonist (IC50 = 10.2 μM). Selectively binds α3β4 over α4β2 and α7 subtypes (Ki values are 0.508, >100 and >100 μM, respectively).

Chemical Identifiers

CAS 1150153-86-8
Molecular Formula C17H22N2O

Specifications

Quantity 10 mg
Formula Weight 270.37
Purity >98%
Molecular Weight (g/mol) 270.376
Chemical Name or Material SR 16584
Recommended Storage Store at -20°C