accessibility menu, dialog, popup

UserName

CH3NNO

CAS RN 1150153-86-8

IUPAC Name: 1-{9-methyl-9-azabicyclo[3.3.1]nonan-3-yl}-2,3-dihydro-1H-indol-2-one
Synonyms: 1-{9-methyl-9-azabicyclo[3.3.1]nonan-3-yl}-3H-indol-2-one
Molecular Weight (g/mol): 270.38
Molecular Formula: C17H22N2O
InChi Key: GIDZCNCCCWFCIN-UHFFFAOYNA-N
SMILES: CN1C2CCCC1CC(C2)N1C(=O)CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results