missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Squalane 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -38°C |
| Color | Incolore |
| Boiling Point | 350°C |
| Quantity | 25 mL |
| Formula Weight | 422.83 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Squalane |
Sécurité et manipulation
| EINECSNumber | (9)-1317 |
| RTECSNumber | XB6070000 |
| TSCA | Yes |