missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Squalane 98.0+%, TCI America™
Supplier: TCI America H0096100ML
| Quantity | 100 mL |
|---|
Chemical Identifiers
| C30H62 | |
| MFCD00008953 | |
| squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane | |
| 2,6,10,15,19,23-hexamethyltetracosane |
Specifications
| Squalane | |
| -38°C | |
| 350°C | |
| 100 mL | |
| squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane | |
| CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C | |
| 422.826 | |
| 422.83 | |
| Liquid |
| 111-01-3 | |
| Colorless | |
| C30H62 | |
| MFCD00008953 | |
| PRAKJMSDJKAYCZ-UHFFFAOYSA-N | |
| 2,6,10,15,19,23-hexamethyltetracosane | |
| 8089 | |
| ≥98.0% (GC) |
Safety and Handling
EINECSNumber : (9)-1317
RTECSNumber : XB6070000
TSCA : Yes