Learn More
Saint-Gobain Silicone Tubing, Platinum-Cured, 0.063 in ID, 0.188 in OD
Numéro de catalogue. NC2389613
Encompass
Sani-Tech SPT-60L is a platinum-cured silicone peristaltic pump tubing engineered for precise pharmaceutical fluid transfer. It is optimized for final-fill and extended-duration pumping where low spallation, long pump life, and accurate dispensing are required.
  • Platinum-cured silicone for pharmaceutical-grade purity.
  • 0.063 in (1.6 mm) inner diameter, 0.188 in (4.8 mm) outer diameter.
  • Wall thickness 0.063 in (1.6 mm) for consistent peristaltic performance.
  • Typical pump life over 250 hours to reduce downtime.
  • Low spallation for cleaner final-fill dispensing.
  • Sterilizable by autoclave, gamma irradiation (up to 5.0 Mrad/50 kGy), or ethylene oxide.
  • Full extractables report and validation package available.
Numéro de catalogue. NC2389613
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Sani-Tech SPT-60L is a platinum-cured silicone peristaltic pump tubing engineered for precise pharmaceutical fluid transfer. It is optimized for final-fill and extended-duration pumping where low spallation, long pump life, and accurate dispensing are required.
  • Platinum-cured silicone for pharmaceutical-grade purity.
  • 0.063 in (1.6 mm) inner diameter, 0.188 in (4.8 mm) outer diameter.
  • Wall thickness 0.063 in (1.6 mm) for consistent peristaltic performance.
  • Typical pump life over 250 hours to reduce downtime.
  • Low spallation for cleaner final-fill dispensing.
  • Sterilizable by autoclave, gamma irradiation (up to 5.0 Mrad/50 kGy), or ethylene oxide.
  • Full extractables report and validation package available.