missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPRY4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA514349
Description
SPRY4 Polyclonal Antibody for Western Blot
SPRY4 is an inhibitor of the receptor-transduced mitogen-activated protein kinase (MAPK) signaling pathway. The protein is positioned upstream of RAS activation and impairs the formation of active GTP-RAS.Identifiants chimiques
| SPRY4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| SPRY4 | |
| A030006O18Rik; HH17; Protein sprouty homolog 4; Sprouty 4; sprouty homolog 4; sprouty homolog 4 (Drosophila); sprouty RTK signaling antagonist 4; sprouty4; Spry4; spry-4 | |
| SPRY4 | |
| Primary | |
| Antigen Affinity Chromatography, Protein A |
| 0.27 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9C004 | |
| 81848 | |
| KLH conjugated synthetic peptide between 270-301 amino acids from the C-terminal region of human SPRY4. | |
| Antibody |
Spécifications
| SPRY4 | |
| Polyclonal | |
| Unconjugated | |
| SPRY4 | |
| A030006O18Rik; HH17; Protein sprouty homolog 4; Sprouty 4; sprouty homolog 4; sprouty homolog 4 (Drosophila); sprouty RTK signaling antagonist 4; sprouty4; Spry4; spry-4 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 81848 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9C004 | |
| SPRY4 | |
| KLH conjugated synthetic peptide between 270-301 amino acids from the C-terminal region of human SPRY4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |