Learn More
Beckman Coulter SPRIselect, 5 mL
Supplier: Beckman Coulter B23317
SPRIselect 5 mL reagent kit is a SPRI-based chemistry that speeds and simplifies nucleic acid size selection for fragment library preparation for next generation sequencing (NGS). SPRIselect yields consistent size selections without bead calibration between reagent lots. Suitable for manual or automated workflows - the SPRIselect method can be run on a variety of Biomek platforms that offer minimal hands-on time.