Learn More
Neogen Corporation Sponge-Stick, biocide-free cellulose sponge, 10 mL neutralizing buffer, gamma irradiated, case of 100, 1.5 x 3 in sponge, 6 x 11 in sample bag, 30 oz capacity, for surface microbial testing
Numéro de catalogue. NC3072440
Encompass
Sponge-Stick, biocide-free cellulose sponge, 10 mL neutralizing buffer, gamma irradiated, case of 100, 1.5 x 3 in sponge, 6 x 11 in sample bag, 30 oz capacity, for surface microbial testing
Numéro de catalogue. NC3072440 Fournisseur Neogen Corporation Code fournisseur NGNSSL10NB
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Sponge-Stick, biocide-free cellulose sponge, 10 mL neutralizing buffer, gamma irradiated, case of 100, 1.5 x 3 in sponge, 6 x 11 in sample bag, 30 oz capacity, for surface microbial testing