Learn More
MilliporeSigma™ Supelco™ SPME Fiber Holder
Description
An adjustable depth guide positions the fiber for sampling and for correct placement in the heated zone of the GC injection port. The fiber can be locked in the exposed position.
Spécifications
Spécifications
| For Use With (Application) | Food and Beverages |
| For Use With (Equipment) | CTC CombiPAL, Gerstel MPS2 and Thermo TriPlus Autosamplers |
| Type | SPME Fibers and Accessories |