missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Splitomicin 98.0+%, TCI America™

$245.58 - $835.28

Chemical Identifiers

CAS 5690-03-9
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD08705254
InChI Key ISFPDBUKMJDAJH-UHFFFAOYSA-N
Synonym 1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one
PubChem CID 5269
ChEBI CHEBI:75272
IUPAC Name 1,2-dihydrobenzo[f]chromen-3-one
SMILES C1CC(=O)OC2=C1C3=CC=CC=C3C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S0892200MG
View Documents
Tci America
S0892200MG
200mg Each for $245.58
 
S08921G
View Documents
Tci America
S08921G
1g Each for $835.28
 
Specifications

Chemical Identifiers

5690-03-9
198.22
ISFPDBUKMJDAJH-UHFFFAOYSA-N
5269
1,2-dihydrobenzo[f]chromen-3-one
C13H10O2
MFCD08705254
1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one
CHEBI:75272
C1CC(=O)OC2=C1C3=CC=CC=C3C=C2

Specifications

5690-03-9
MFCD08705254
ISFPDBUKMJDAJH-UHFFFAOYSA-N
1,2-dihydrobenzo[f]chromen-3-one
5269
198.22
≥98.0% (GC)
55°C
Splitomicin
C13H10O2
1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one
C1CC(=O)OC2=C1C3=CC=CC=C3C=C2
198.22
CHEBI:75272
Crystal-Powder at 20°C
White
200mg
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator