missing translation for 'onlineSavingsMsg'
Learn More

Splitomicin 98.0+%, TCI America™

Supplier:  TCI America S08921G

 View more versions of this product

Catalog No. S08921G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Splitomicin
Crystalline Powder
55°C
1 g
MFCD08705254
ISFPDBUKMJDAJH-UHFFFAOYSA-N
1,2-dihydrobenzo[f]chromen-3-one
5269
198.22
Splitomicin
5690-03-9
White
C13H10O2
1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one
C1CC(=O)OC2=C1C3=CC=CC=C3C=C2
198.22
CHEBI:75272
≥98.0% (GC)

Chemical Identifiers

5690-03-9
198.22
ISFPDBUKMJDAJH-UHFFFAOYSA-N
5269
1,2-dihydrobenzo[f]chromen-3-one
C13H10O2
MFCD08705254
1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one
CHEBI:75272
C1CC(=O)OC2=C1C3=CC=CC=C3C=C2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions