missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Splitomicin 98.0+%, TCI America™
Supplier: TCI America S0892200MG
Specifications
| Splitomicin | |
| Crystalline Powder | |
| 55°C | |
| 200 mg | |
| MFCD08705254 | |
| ISFPDBUKMJDAJH-UHFFFAOYSA-N | |
| 1,2-dihydrobenzo[f]chromen-3-one | |
| 5269 | |
| 198.22 |
| Splitomicin | |
| 5690-03-9 | |
| White | |
| C13H10O2 | |
| 1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one | |
| C1CC(=O)OC2=C1C3=CC=CC=C3C=C2 | |
| 198.22 | |
| CHEBI:75272 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 5690-03-9 | |
| 198.22 | |
| ISFPDBUKMJDAJH-UHFFFAOYSA-N | |
| 5269 | |
| 1,2-dihydrobenzo[f]chromen-3-one |
| C13H10O2 | |
| MFCD08705254 | |
| 1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one | |
| CHEBI:75272 | |
| C1CC(=O)OC2=C1C3=CC=CC=C3C=C2 |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator