missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SPINeasy™ DNA Kit for Plant
Description
- SPINeasy DNA Kit for Plant is a silica-membrane spin-column kit that enables quick, convenient and cost-effective isolation of genomic DNA (gDNA) from various plant tissues, such as leaf, stem and seed.
- High quality gDNA can be extracted from polysaccharide and polyphenolic-rich plants.
- The use of our specially formulated Lysis Buffer PD and Lysing Matrix A in combination with FastPrep™ Instruments from MP Biomedicals enables highly efficient lysis of various types of samples within seconds.
- For DNA purification, Column PD and kit buffers are designed to deliver gDNA extracts of high yield and purity which are compatible with downstream molecular biology applications such as PCR, restriction digestion and sequencing.
Specifications
Specifications
| Content And Storage | Store at Room Temperature, Dry Environment |
| Format | Kit |
| Isolation Technology | Spin Column |
| Sample Type | Plant Tissues, 50 to 150 mg |
| Includes | 50 x 2mL Lysing Matrix A, 50 mL Lysis Buffer PD, 12 mL PPS Solution, 15 mL Binding Buffer PD, 2 x 12 mL Wash Buffer, 10 mL Elution Buffer PD, 0.8 mL RNase A, 50 x Column PD with collection tubes. |
| Product Type | gDNA Extraction Kit |
| Quantity | 50 Preps |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.