Learn More
New Pig Spill Kit in High - Visibility Economy Container
High-visibility, low-cost PIG Spill Kit is perfect for everyday spill response. New Pig best kit for everyday spills is packed with top-quality PIG Absorbents
Supplier:  New Pig KIT2200
Catalog No. 17987834
Only null left
Description
Description
  • Highly visible for quick response: The bright yellow container is easy to spot in an emergency, ensuring your spill supplies can be located quickly when every second counts.
  • Affordable for widespread use: Its economical design makes it cost-effective to stock in multiple spill-prone areas, ensuring a fast response no matter where a leak or spill occurs.
  • Packed with top-quality PIG Absorbents: Despite its low cost, this kit contains the same high-quality PIG Absorbents that are trusted for rapid and effective spill cleanup.
  • Universal absorbency: The kit is designed to absorb a variety of common liquids, including oils, coolants, solvents, and water, providing a versatile solution for everyday spills.
  • Basic, easy-to-use container: The simple, non-threaded lid lifts off for fast access to supplies, and the container is made of a lightweight and chemical-resistant polyethylene.
  • Ideal for everyday spills: This kit is the perfect solution for routine spills and leaks, providing a quick, simple, and effective way to maintain a clean and safe workplace.
Product Suggestions
Product Suggestions