Learn More
FyterTech Nonwovens Spilfyter™ Universal Spill Containment Products
Socks, pillows and drum tops for acids or bases, and nonagressive spills, such as oil, coolants, solvents, and water
$74.08 - $165.21
Products 10
Catalog Number Mfr. No. Dimensions (L x W) Type Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Type Packaging Quantity Price Quantity  
19-066-573
View Documents
FyterTech Nonwovens
G61
10 x 10 in. Universal Sorbent Pillow 20/Cs.
Case of 20 Each for $97.82
Only null left
19-066-574
View Documents
FyterTech Nonwovens
G62
18 x 18 in. Universal Sorbent Pillow 10/Cs.
Case of 10 Each for $120.48
Only null left
19-066-562
View Documents
FyterTech Nonwovens
DB75
18 x 16 in. Universal Sorbent Pad 100/Cs.
Case of 100 Each for $120.89
Only null left
19-122-885
View Documents
FyterTech Nonwovens
G66
17 x 8.5 in. Universal Sorbent Pillow 10/Cs.
Case of 10 Each for $85.00
Only null left
19-066-572
View Documents
FyterTech Nonwovens
G37
10 ft. x 3 in. Universal Sorbent Sock 6/Cs.
Case of 6 Each for $78.64
Only null left
19-067-800
View Documents
FyterTech Nonwovens
37031080
20 x 16 in. Light-duty Soaker Roll 250/Cs.
Case of 250 Each for $156.46
Only null left
19-066-571
View Documents
FyterTech Nonwovens
G34
4 ft. x 3 in. Universal Sorbent Sock 40/Cs.
Case of 40 Each for $155.24
Only null left
19-066-578
View Documents
FyterTech Nonwovens
G76
Universal Sorbent Drum Top 25/Cs.
Case of 25 Each for $96.93
Only null left
19-066-570
View Documents
FyterTech Nonwovens
G30
4 ft. x 3 in. Universal Sorbent Sock 15/Cs.
Case of 15 Each for $74.08
Only null left
19-123-621
View Documents
FyterTech Nonwovens
47031018
20 x 16 in. Heavy-duty Soaker Pad 200/Cs.
Case of 200 Each for $165.21
Only null left
Description
Description
Use with Spilfyter pads and rolls for complete coverage.
Socks
  • Contain polypropylene for superior absorption, plus polyester casing for excellent stretching ability
  • Surround spill with socks to prevent spreading or to use as a barricade
  • Absorb and retain oil, water, solvents, coolants, etc, to prevent spreading spills, to wrap around leaky drums, tanks, sinks, machine bases for leak protection, useful as a barricade, makes cleanup quick and easy
Pillows
  • Pillows soak up different size spills in ponds, ditches, open water or under machinery
  • Pillow-in-a-Pan Kit holds liquids within plastic pan that also holds pillow
  • Absorb and retain oil, water, solvents, coolants and many more, contain spills and prevent migration, makes cleanup quick and easy
Drum Tops
  • Precut hole provides a perfect fit around pump
  • Superior absorbency keeps drum tops clean
  • Absorb and retain oil, water, solvents, coolants, etc., place on drum tops to soak up pump drips and keep work areas neat and clean
Soaker Roll/Pad
  • Provides absorption protection from spills and cushioning to minimize breakage
  • Sorbent Pad
    • Absorb and retain oil, water, solvents, coolants and many more, used as a rag for quick wipe ups, use to line shelves and for clean-up applications
  • Spill Kit
    • Complete spill response capability for either inside or outside storage
    Videos
    SDS
    missing translation for 'documents'
    missing translation for 'documents'