missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FyterTech Nonwovens Spilfyter™ KolorSafe™ Neutralizers for Acids
Description
Unlike dry neutralizer, liquid reagent can be sprayed into hard-to-reach areas and on vertical surfaces.
- Sodium-free liquid reagent changes color when neutralization is complete
- Safe for use in laboratories and cleanrooms
Specifications
Specifications
| Capacity (English) | 1 gal. |
| Capacity (Metric) | 3.78 L |
| Type | Acid Neutralizer |
| Format | Liquid |
| For Use With (Application) | Spill control |
| Includes | Color change indicator when neutralized |
| Packaging Quantity | 4/Cs. |
| Weight (English) | 38 lb. |
| Weight (Metric) | 17 kg |