missing translation for 'onlineSavingsMsg'
Learn More

FyterTech Nonwovens Spilfyter™ Granular Sorbents

Designed for efficient cleaning of oils and grease

Supplier:  FyterTech Nonwovens JV 888002

Numéro de catalogue. 296351848

Il en reste null

Description

Description

  • All-purpose oil and grease absorbent
  • 1:1 weight to weight absorbency ratio
  • Components: Paper fiber; Kaolin; Calcium carbonate; Titanium dioxide

Order Info

Packaged in 14kg (30 lb.) bag

Spécifications

Spécifications

Sorbent
Efficient cleaning of oils and grease
30 lb.
Granular
30 lb.
14 kg
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions