Learn More
Abcam Inc Spermine tetrahydrochloride, polyamine used for precipitation of DNA, 5G
Numéro de catalogue. 502704598
Encompass
MW 348.19 g/mol. Spermine is a biogenic polyamine formed from spermidine and occurs in almost all tissues. It is essential for both neoplastic and normal tissue growth. Spermine is associated with nucleic acids, particularly in viruses, and is thought to stabilize the helical structure. Spermine tetrahydrochloride is used for precipitation of DNA from low salt aqueous buffers.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502704598 Fournisseur Abcam Inc Code fournisseur AB1465235G
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 348.19 g/mol. Spermine is a biogenic polyamine formed from spermidine and occurs in almost all tissues. It is essential for both neoplastic and normal tissue growth. Spermine is associated with nucleic acids, particularly in viruses, and is thought to stabilize the helical structure. Spermine tetrahydrochloride is used for precipitation of DNA from low salt aqueous buffers.
The product is subject to the following: Abcam Restricted Use Statement