missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Spectrum Tubes

For demonstrating spectral lines of pure substances.

Supplier:  Eisco™ PH1196B10

 Afficher plus de versions de ce produit

Numéro de catalogue. S13005


Il en reste null

Comprend:

Spectrum Tube


Description

Description

  • Features large 10cm capillary in center for greater viewing area
  • For use with power supplies holding 26cm spectrum tubes
  • Designed for operation intervals of 30 seconds
  • Should be used for 30 seconds on/30 seconds off
Spécifications

Spécifications

Spectrum Tube, Xenon
Demonstrates the spectral lines of pure substances
Spectrum Tube, large 10cm capillary
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions