missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Transfertube™ Disposable Sampling Pipet and Dropper

Catalog No. 137112
Change view
Click to view available options
Volume (Metric):
4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
137112 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 137112 Supplier Spectrum™ Supplier No. 190195
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An innovative device for transferring liquid and coring semi-solid samples with one hand

The large aperture is particularly useful for working with particulates, suspensions, pastes, gels & creams. Disposable transferring reduces the risk of cross contamination associated with re-used pipettes and cutting instruments.

  • Capacity: 4mL
  • Length: 6.8 in. (175mm)
  • Shaft and plunger L x Dia.: 6 x 0.24 in. (153 x 6mm)
  • Finger ring I.D. x O.D.: 0.65 x 0.8 in. (16.5 x 20mm)
Applications:
  • General: Powders, viscous solutions, slurries, liquids
  • Microbiology: bacterial or viral colony on agar
  • Ecology and Industrial: Soil, mud, sludge, core samples, oils
  • Cosmetics: creams, gels, waxes, sampling
  • Food processing: pastes, creams, syrups, concentrates

Specifications

Material Polypropylene
For Use With (Application) For bacterial or viral colony on agar in microbiology, powders, viscous solutions, slurries, liquids, soil, mud, sludge, core samples
Length (English) 6 in.
Volume (Metric) 4 mL
Length (Metric) 15.24 cm
Type Transfer Pipet
Disposable Single-use