missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Specimen Chucks for QS and Microm Cryostats, 30 mm

Catalog No. 22445834
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Circular Chuck
Waffle Chuck
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
22445834 Yellow Circular Chuck
22445694 Blue Circular Chuck
22445307 Green Circular Chuck
22445496 Orange Circular Chuck
22445574 Pink Circular Chuck
22445720 Purple Circular Chuck
22445500 Red Circular Chuck
22445504 White Circular Chuck
22445627 Blue Waffle Chuck
22445629 Green Waffle Chuck
22445576 Orange Waffle Chuck
22445456 Pink Waffle Chuck
22445458 Purple Waffle Chuck
22445686 Red Waffle Chuck
22445474 White Waffle Chuck
22445688 Yellow Waffle Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 22445834 Supplier Mopec™ Supplier No. SP133YE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Specifications

Product Type Circular Chuck
For Use With (Equipment) QS and Microm Cryostats
Color Yellow