missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Specimen Chucks for CRYOSTAR Cryostats, 40 mm

Numéro de catalogue. p-200007554
Change view
Click to view available options
Color:
Blue
Green
Orange
Pink
Purple
Red
White
Yellow
Product Type:
Circular Chuck
Waffle Chuck
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Color Product Type
22445306 Blue Circular Chuck
22445477 Green Circular Chuck
22445497 Orange Circular Chuck
22445475 Pink Circular Chuck
22445626 Purple Circular Chuck
22445327 Red Circular Chuck
22445556 White Circular Chuck
22445575 Yellow Circular Chuck
22445557 Blue Waffle Chuck
22445472 Green Waffle Chuck
22445333 Orange Waffle Chuck
22445658 Pink Waffle Chuck
22445495 Purple Waffle Chuck
22445697 Red Waffle Chuck
22445480 White Waffle Chuck
22445313 Yellow Waffle Chuck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Numéro de catalogue. 22445306 Fournisseur Mopec™ Code fournisseur SP124BL
Il en reste null

Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.

  • Round, aluminum specimen chucks

Spécifications

Product Type Circular Chuck
For Use With (Equipment) CryoStar NX50 and NX70
Material Aluminum
Color Blue