Learn More
Fisherbrand™ Specific Gravity/Baume Hydrometers
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
Description
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
- Permanently marked with individual serial numbers
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
Certificate of Accuracy and Compliance provided to indicate calibration against standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications
Spécifications
| Specific Gravity Hydrometer | |
| 3-point Cert, NIST, ASTM E100, E126 | |
| 300mL Borosilicate Glass cylinder | |
| 1 to 2 |
| 0.02 | |
| 165 mm | |
| Measure the specific gravity of liquids heavier than Water | |
| 140 mL |