Learn More
Fisherbrand™ Specific Gravity/Baume Hydrometers
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
 Afficher plus de versions de ce produit
Numéro de catalogue. 11540E
Il en reste null
Description
Description
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
  • Permanently marked with individual serial numbers
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
Certificate of Accuracy and Compliance provided to indicate calibration against standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications
Spécifications
Spécifications
Specific Gravity Hydrometer
NIST, ASTM E100, ASTM E126
300mL Borosilicate Glass cylinder
Measure the specific gravity of liquids heavier than Water
250 mL
0.01
285 mm
Soda Lime Glass
1.000 to 2.000
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Accessoires