missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC HandySorb™ Mop System and Supplies

Numéro de catalogue. 19129308
Change view
Click to view available options
Product Type:
Absorbent Pads, Mop Adapter
Absorbent Pillows
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Product Type
19129308 Absorbent Pillows
19129307 Absorbent Pads, Mop Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 19129308 Fournisseur Brady™ Code fournisseur HANDYSORBNTPILLOW
Il en reste null

SPC HandySorb Mop System™ is ideal for cleaning hard-to-reach spills. Eliminates kneeling and gets the job done in three simple steps – snap on, absorb and release.

  • Keep hands out of the mess with the No-Touch Universal Pads and Pillows that clean up a variety of fluids
  • Increase reach – handle extends to 6 feet
  • Patented quick-release operation makes disposal of used pads simple and keeps hands clean.

Spécifications

Product Type Absorbent Pillows
Color Gray
Sterility Non-sterile
Autoclavable Not Autoclavable
Width (English) 18 in.
Quantity 10
For Use With (Application) HandySorb Mop
Material Polypropylene