Learn More
Brady™ Spare LED Bulb for Line Projector - 80 W/150 W
Restores original projector brightness for optimal visibility.
$584.44
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
|
17823779
|
Brady™
178988 |
Blue |
Each for $584.44
|
|
|||||
|
17823780
|
Brady™
178989 |
Green |
Each for $584.44
|
|
|||||
|
17823781
|
Brady™
178990 |
Red |
Each for $584.44
|
|
|||||
|
17823782
|
Brady™
178991 |
Yellow |
Each for $584.44
|
|
|||||
Description
- Specifically designed to replace the existing LED in compatible projectors.
- Copper mounting plate ensures efficient heat transfer for optimal LED performance and lifespan.
- Replacement of the LED requires soldering and basic electrical skills and should be performed by a qualified electrician.
- LED can only be operated when mounted onto the projector heat sink otherwise damage can occur.
Specifications
| Bulb | |
| LED |