Learn More
Brady™ Spare LED Bulb for Line Projector - 80 W/150 W
Restores original projector brightness for optimal visibility.
Supplier: Brady™ 178989
Description
- Specifically designed to replace the existing LED in compatible projectors.
- Copper mounting plate ensures efficient heat transfer for optimal LED performance and lifespan.
- Replacement of the LED requires soldering and basic electrical skills and should be performed by a qualified electrician.
- LED can only be operated when mounted onto the projector heat sink otherwise damage can occur.
Chemical Identifiers
| Bulb | |
| LED |
Specifications
| Bulb | |
| LED | |
| Green |