Learn More
Brady™ Spare LED Bulb for Line Projector - 80 W/150 W
Restores original projector brightness for optimal visibility.
Supplier:  Brady™ 178990
 View more versions of this product
Catalog No. 17823781
Only null left
Description
Description
  • Specifically designed to replace the existing LED in compatible projectors.
  • Copper mounting plate ensures efficient heat transfer for optimal LED performance and lifespan.
  • Replacement of the LED requires soldering and basic electrical skills and should be performed by a qualified electrician.
  • LED can only be operated when mounted onto the projector heat sink otherwise damage can occur.
Specifications
Chemical Identifiers
Bulb
LED
Specifications
Bulb
LED
Red
Product Suggestions
Product Suggestions