Learn More
Brady™ Spare LED Bulb for Line Projector - 80 W/150 W
Description
- Specifically designed to replace the existing LED in compatible projectors.
- Copper mounting plate ensures efficient heat transfer for optimal LED performance and lifespan.
- Replacement of the LED requires soldering and basic electrical skills and should be performed by a qualified electrician.
- LED can only be operated when mounted onto the projector heat sink otherwise damage can occur.
Specifications
Specifications
| Product Type | Bulb |
| Display | LED |
| Color | Yellow |