missing translation for 'onlineSavingsMsg'
Learn More

Span 80 (=Sorbitan Monooleate), TCI America™

Catalog No. S0060500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0060500G 500 g
S006025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0060500G Supplier TCI America Supplier No. S0060500G

Chemical Identifiers

CAS 1338-43-8
Molecular Formula C24H44O6
Molecular Weight (g/mol) 428.61
MDL Number MFCD00080948
InChI Key NWGKJDSIEKMTRX-NZAQQJATSA-N
Synonym Arlacel 80, Sorbitan Monooleate
PubChem CID 70693544
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

Color Brown-Yellow
Physical Form Clear Liquid at 20°C
Quantity 500 g
Chemical Name or Material Span 80 (=Sorbitan Monooleate)
EINECSNumber (8)-0063
RTECSNumber WG2932400
TSCA Yes