missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Pipette Pump™ Pipetter

Catalog No. S35821
Change view
Click to view available options
Color:
Green
Volume (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric)
S35821 Green 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S35821 Supplier Bel-Art™ Supplier No. F378980000/EMD

Provides safe, accurate. and trouble-free pipetting. Pipetters utilize an easy-to-use one-handed operation.

  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121°C (250°F) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available

Specifications

Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Green
Disposable Reusable
For Use With Serologic Pipette
Grip Type Ergonomic
Material Polypropylene
Model Pipette Pump Pipettors
Number of Channels 1
Sterility Non-sterile
Type Pipette
Volume (Metric) 10 mL
For Use With (Equipment) Pipettes
Resistance Acid and Alkali
Show More Show Less