missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Pipet Filler

Catalog No. S43303
Change view
Click to view available options
Number of Channels:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
S43303 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43303 Supplier Bel-Art™ Supplier No. F378860000/EMD
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Squeeze, then release the bulb to draw fluid into the pipet

  • The pipet aid is removed when fluid fills the bulb, and the pipet top quickly covered with a gloved finger to safely and easily control and release fluid
  • The bulb is Vikem™ vinyl with a tapered polyethylene chuck for quick removal when pipet is filled

Specifications

Autoclavable Not Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette
Grip Type Round
Material Vinyl
Model Standard
Number of Channels 1
Sterility Non-sterile
Type Pipette Filler
For Use With (Equipment) Pipet
Show More Show Less