missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ LabMat™ Bench Liners
Description
- Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
- Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
- Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
- Roll up and discard in proper waste container after contamination
- Select orange to identify use with biohazard materials
Specifications
Specifications
| Product Type | Bench Protector |
| Material | Polyethylene |
| Absorbancy | 400 mL/m2 |
| Length (English) | 24 in. |
| Length (Metric) | 610 mm |
| Width (English) | 20 in. |
| Width (Metric) | 508 mm |
| Dimensions (L x W) | 24 x 20 in. (610 x 508mm) |
| Color | White |
| Description | Pack of 100 sheets |
| Show More |