missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Plastic Tubing Cutter and Replacement Blades
Description
- Spring-loaded jaw will remain closed for safety when not in use
- Replaceable, hardened steel blade makes clean, straight cuts
Spécifications
Spécifications
| Material | Plastic |
| Overall Length (English) | 3 in. |
| Overall Length (Metric) | 76.2 mm |
| For Use With | 1.6 to 19mm Tubing |
| Product Type | Tubing Cutter |
Not recommended for latex tubing.