missing translation for 'onlineSavingsMsg'
Learn More

Soybean Oil, USP, Spectrum™ Chemical

Catalog No. 18606967
Change view
Click to view available options
Quantity:
20 L
200 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606967 200 L Amber Glass Bottle
18606966 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606967 Supplier Spectrum Chemical Mfg Cor Supplier No. SO255200LTBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO255, 8001-22-7

Spectrum™ Chemical Soybean Oil, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8001-22-7

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 200 L
Grade USP
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Soybean Oil