missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitol Solution, USP, 70%, Spectrum™ Chemical
SO220, 50-70-4, C6H14O6
$1,098.15
Chemical Identifiers
| CAS | 50-70-4 , 7732-18-5 |
|---|---|
| Molecular Formula | C6H14O6 |
| Molecular Weight (g/mol) | 182.17 |
| InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
| IUPAC Name | hexane-1,2,3,4,5,6-hexol |
| SMILES | OCC(O)C(O)C(O)C(O)CO |